Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnpy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077585.1 | NP_001071053.1 | 5 - 6 | 823 | 69 | |
GenBank mRNA | BC126101.1 | - | 5 - 6 | 823 | 7319422 | 69 |
FQ220100.1 | - | 5 - 6 | 679 | 69 | ||
FQ225671.1 | - | 5 - 6 | 759 | 69 | ||
FQ231871.1 | - | 5 - 6 | 730 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |