Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079936.1 | NP_001073405.1 | 4 - 5 | 705 | 115 | |
XM_006240750.2 | XP_006240812.1 | 4 - 5 | 628 | 115 | ||
XM_006240751.2 | XP_006240813.1 | 5 - 6 | 726 | 115 | ||
XM_006240752.2 | XP_006240814.1 | 4 - 5 | 853 | 115 | ||
GenBank mRNA | BC129101.1 | - | 4 - 5 | 705 | 7098463 | 115 |
FQ211489.1 | - | 4 - 5 | 843 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |