Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Luc7l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107853.1 | NP_001101323.1 | 8 - 9 | 1151 | 70 | |
XM_006236314.2 | XP_006236376.1 | 10 - 11 | 990 | 70 | ||
XM_006236315.2 | XP_006236377.1 | 9 - 10 | 1654 | 70 | ||
XM_006236316.2 | XP_006236378.1 | 9 - 10 | 883 | 70 | ||
XM_006236319.2 | XP_006236381.1 | 8 - 9 | 1583 | 70 | ||
XM_006236322.2 | XP_006236384.1 | 8 - 9 | 836 | 70 | ||
XM_006236323.2 | XP_006236385.1 | 10 - 11 | 1352 | 70 | ||
XM_008762778.1 | XP_008761000.1 | 9 - 10 | 3790 | 70 | ||
XM_008762780.1 | XP_008761002.1 | 9 - 10 | 1090 | 70 | ||
XM_008762781.1 | XP_008761003.1 | 8 - 9 | 1603 | 70 | ||
GenBank mRNA | BC166855.1 | - | 8 - 9 | 1172 | 9099059 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |