Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arih1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013108.3 | NP_001013126.2 | 11 - 12 | 1479 | 104 | |
GenBank mRNA | AW141789.1 | - | 1 - 2 | 66 | 104 | |
BC083881.1 | - | 9 - 10 | 1364 | 7131418 | 104 | |
DN956151.1 | - | 8 - 9 | 641 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |