Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ryr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008759293.1 | XP_008757515.1 | 85 - 86 | 12364 | 92 | |
GenBank mRNA | AF130879.1 | - | 2 - 3 | 157 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Ion Channel Detect Genomic DNA Rattus norvegicus Probe spans exons |