Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scamp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023955.1 | NP_076445.1 | 6 - 7 | 732 | 116 | |
GenBank mRNA | AF295405.1 | - | 5 - 6 | 576 | 116 | |
BC085862.1 | - | 6 - 7 | 732 | 7190371 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |