Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlgap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276304.1 | NP_001263233.1 | 4 - 5 | 1513 | 109 | |
XM_008763989.1 | XP_008762211.1 | 5 - 6 | 2815 | 109 | ||
GenBank mRNA | AY530298.1 | - | 3 - 4 | 1376 | 109 | |
AY530299.1 | - | 3 - 4 | 1376 | 109 | ||
U67139.1 | - | 4 - 5 | 1480 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |