Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcmf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128192.1 | NP_001121664.1 | 2 - 3 | 422 | 93 | |
GenBank mRNA | BC167075.1 | - | 2 - 3 | 422 | 9093013 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |