Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Retsat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145084.1 | NP_659552.1 | 2 - 3 | 347 | 77 | |
GenBank mRNA | AF465614.1 | - | 2 - 3 | 347 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |