Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ing4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079887.1 | NP_001073356.1 | 2 - 3 | 120 | 80 | |
XM_006237349.2 | XP_006237411.1 | 2 - 3 | 219 | 80 | ||
XM_006237350.2 | XP_006237412.1 | 2 - 3 | 219 | 80 | ||
GenBank mRNA | BC086570.1 | - | 2 - 3 | 120 | 80 | |
BC097942.1 | - | 2 - 3 | 121 | 80 | ||
BC127511.1 | - | 2 - 3 | 120 | 7376723 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |