Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grin2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022797.1 | NP_073634.1 | 6 - 7 | 2308 | 57 | |
XM_008759316.1 | XP_008757538.1 | 5 - 6 | 2013 | 57 | ||
XM_008759317.1 | XP_008757539.1 | 5 - 6 | 2013 | 57 | ||
GenBank mRNA | D13213.1 | - | 5 - 6 | 1602 | 57 | |
L31611.1 | - | 5 - 6 | 1583 | 57 | ||
L31612.1 | - | 6 - 7 | 2308 | 57 | ||
U08260.1 | - | 6 - 7 | 1654 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Detect Genomic DNA Rattus norvegicus Probe spans exons |