Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnq3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031597.4 | NP_113785.3 | 13 - 14 | 2223 | 72 | |
XM_008765495.1 | XP_008763717.1 | 13 - 14 | 1551 | 72 | ||
GenBank mRNA | AF091247.1 | - | 14 - 15 | 1769 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Rattus norvegicus Probe spans exons |