Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106442.1 | NP_001099912.1 | 5 - 6 | 525 | 67 | |
GenBank mRNA | BC168754.1 | - | 5 - 6 | 497 | 6886830 | 67 |
FQ213377.1 | - | 5 - 6 | 488 | 66 | ||
FQ221799.1 | - | 5 - 6 | 463 | 67 | ||
FQ226826.1 | - | 5 - 6 | 463 | 67 | ||
FQ229540.1 | - | 5 - 6 | 497 | 67 | ||
FQ232929.1 | - | 5 - 6 | 481 | 67 | ||
FQ235190.1 | - | 5 - 6 | 481 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |