Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbl1x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106964.1 | NP_001100434.1 | 1195 | 70 | ||
GenBank mRNA | BC167084.1 | - | 1290 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |