Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138911.3 | NP_620266.1 | 1 - 2 | 71 | 82 | |
GenBank mRNA | BC061529.1 | - | 1 - 2 | 72 | 6887497 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |