Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110793.1 | NP_001104263.1 | 10 - 11 | 1736 | 63 | |
NM_058210.2 | NP_478117.2 | 10 - 11 | 1736 | 63 | ||
XM_006230641.2 | XP_006230703.1 | 10 - 11 | 4637 | 63 | ||
XM_006230642.2 | XP_006230704.1 | 10 - 11 | 4616 | 63 | ||
XM_006230646.2 | XP_006230708.1 | 9 - 10 | 4561 | 63 | ||
XM_008760065.1 | XP_008758287.1 | 8 - 9 | 2550 | 63 | ||
XM_008760066.1 | XP_008758288.1 | 10 - 11 | 4637 | 63 | ||
GenBank mRNA | BC081859.1 | - | 9 - 10 | 1649 | 7132476 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |