Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271381.1 | NP_001258310.1 | 3 - 4 | 156 | 77 | |
XM_008772413.1 | XP_008770635.1 | 2 - 3 | 321 | 77 | ||
GenBank mRNA | BC168970.1 | - | 2 - 3 | 322 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |