Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012057.1 | NP_001012057.1 | 1 - 2 | 268 | 64 | |
XM_008772405.1 | XP_008770627.1 | 2 - 3 | 389 | 64 | ||
XM_008772406.1 | XP_008770628.1 | 2 - 3 | 578 | 64 | ||
XM_008772407.1 | XP_008770629.1 | 2 - 3 | 315 | 64 | ||
GenBank mRNA | FQ223845.1 | - | 464 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |