Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001010953.1 | NP_001010953.1 | 2 - 3 | 335 | 62 | |
GenBank mRNA | BC089058.1 | - | 2 - 3 | 335 | 7315480 | 62 |
D82867.1 | - | 2 - 3 | 365 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |