Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lin7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006241305.2 | XP_006241367.1 | 1 - 2 | 55 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |