Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dync1li2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006255053.2 | XP_006255115.1 | 10 - 11 | 1363 | 69 | |
GenBank mRNA | AB008521.1 | - | 5 - 6 | 415 | 69 | |
AB008524.1 | - | 5 - 6 | 415 | 69 | ||
BC089794.1 | - | 10 - 11 | 1170 | 7115029 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |