Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172091.2 | NP_742088.1 | 12 - 13 | 1316 | 96 | |
NM_172092.2 | NP_742089.1 | 12 - 13 | 1324 | 96 | ||
NR_073147.1 | 11 - 12 | 1217 | 96 | |||
XM_006247876.2 | XP_006247938.1 | 12 - 13 | 1430 | 96 | ||
XM_006247877.1 | XP_006247939.1 | 10 - 11 | 1155 | 96 | ||
XM_008768468.1 | XP_008766690.1 | 11 - 12 | 1373 | 96 | ||
GenBank mRNA | BC081700.1 | - | 11 - 12 | 1181 | 7192550 | 96 |
L04796.1 | - | 12 - 13 | 1286 | 96 | ||
X68692.1 | - | 12 - 13 | 1286 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |