Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Habp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001505.1 | NP_001001505.1 | 6 - 7 | 588 | 74 | |
XM_006231631.2 | XP_006231693.1 | 6 - 7 | 636 | 74 | ||
XM_006231632.2 | XP_006231694.1 | 5 - 6 | 525 | 74 | ||
XM_006231633.2 | XP_006231695.1 | 5 - 6 | 522 | 74 | ||
GenBank mRNA | AB177406.1 | - | 6 - 7 | 588 | 74 | |
BC129080.1 | - | 5 - 6 | 474 | 8374257 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |