Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnma1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031828.1 | NP_114016.1 | 1 - 2 | 875 | 100 | |
GenBank mRNA | AF083341.1 | - | 1 - 2 | 311 | 100 | |
AF135265.1 | - | 1 - 2 | 729 | 100 | ||
AY330290.1 | - | 1 - 2 | 200 | 100 | ||
AY330291.1 | - | 1 - 2 | 200 | 100 | ||
AY330292.1 | - | 1 - 2 | 100 | |||
AY330293.2 | - | 1 - 2 | 200 | 100 | ||
AY330294.1 | - | 1 - 2 | 200 | 100 | ||
AY344964.1 | - | 1 - 2 | 200 | 100 | ||
AY344965.2 | - | 1 - 2 | 704 | 100 | ||
U40603.1 | - | 1 - 2 | 875 | 100 | ||
U55995.1 | - | 1 - 2 | 353 | 100 | ||
U93052.1 | - | 1 - 2 | 301 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Rattus norvegicus Probe spans exons |