Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntsr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022695.2 | NP_073186.1 | 2 - 3 | 937 | 86 | |
GenBank mRNA | X97121.1 | - | 2 - 3 | 939 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Rattus norvegicus Probe spans exons |