Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031357.1 | NP_112647.1 | 7 - 8 | 893 | 86 | |
GenBank mRNA | AB043870.1 | - | 7 - 8 | 893 | 86 | |
AY724484.1 | - | 7 - 8 | 894 | 86 | ||
BC081775.1 | - | 7 - 8 | 902 | 7107183 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |