Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif3m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168523.1 | NP_001161995.1 | 9 - 10 | 1003 | 103 | |
GenBank mRNA | FQ220907.1 | - | 9 - 10 | 976 | 103 | |
FQ222771.1 | - | 9 - 10 | 995 | 103 | ||
FQ229546.1 | - | 9 - 10 | 997 | 103 | ||
FQ229811.1 | - | 9 - 10 | 997 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |