Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ivns1abp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047085.1 | NP_001040550.1 | 1 - 2 | 174 | 73 | |
XM_006249988.2 | XP_006250050.1 | 6 - 7 | 787 | 73 | ||
GenBank mRNA | AY553870.2 | - | 1 - 2 | 174 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons |