Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034946.1 | NP_001030118.1 | 4 - 5 | 475 | 70 | |
GenBank mRNA | BC099766.1 | - | 4 - 5 | 475 | 7931002 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |