Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spop | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100496.1 | NP_001093966.1 | 6 - 7 | 816 | 80 | |
XM_006247180.2 | XP_006247242.1 | 7 - 8 | 1620 | 80 | ||
XM_006247181.2 | XP_006247243.1 | 8 - 9 | 927 | 80 | ||
XM_006247182.2 | XP_006247244.1 | 7 - 8 | 876 | 80 | ||
XM_008767968.1 | XP_008766190.1 | 7 - 8 | 972 | 80 | ||
XM_008767969.1 | XP_008766191.1 | 8 - 9 | 894 | 80 | ||
GenBank mRNA | BC091435.1 | - | 4 - 5 | 542 | 80 | |
BC166743.1 | - | 6 - 7 | 816 | 7938440 | 80 | |
FQ217100.1 | - | 8 - 9 | 1124 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |