Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106412.1 | NP_001099882.1 | 4 - 5 | 1026 | 94 | |
XM_006231957.2 | XP_006232019.1 | 5 - 6 | 1127 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Apoptosis 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |