Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubxn2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109482.1 | NP_001102952.1 | 4 - 5 | 471 | 143 | |
XM_006239861.2 | XP_006239923.1 | 4 - 5 | 530 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |