Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039016.2 | NP_001034105.2 | 4 - 5 | 776 | 86 | |
XM_006257521.2 | XP_006257583.1 | 4 - 5 | 878 | 86 | ||
GenBank mRNA | AY886526.1 | - | 4 - 5 | 746 | 86 | |
BC129070.1 | - | 4 - 5 | 776 | 7461054 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |