Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053766.1 | NP_446218.1 | 3 - 4 | 367 | 89 | |
GenBank mRNA | BC089052.1 | - | 3 - 4 | 367 | 7307299 | 89 |
FQ226684.1 | - | 2 - 3 | 149 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |