Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191693.1 | NP_001178622.1 | 3337 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |