Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139261.1 | NP_640354.1 | 7 - 8 | 1120 | 102 | |
XM_006228248.1 | XP_006228310.1 | 7 - 8 | 1347 | 102 | ||
XM_006228249.2 | XP_006228311.1 | 7 - 8 | 1112 | 102 | ||
XM_006228250.2 | XP_006228312.1 | 7 - 8 | 1176 | 102 | ||
GenBank mRNA | AF187860.1 | - | 7 - 8 | 1120 | 102 | |
BC072541.1 | - | 7 - 8 | 1156 | 7103744 | 102 | |
FQ232199.1 | - | 7 - 8 | 1176 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |