Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smek1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108050.2 | NP_001101520.2 | 5 - 6 | 1455 | 86 | |
NM_001271174.1 | NP_001258103.1 | 5 - 6 | 1713 | 86 | ||
GenBank mRNA | FQ087395.1 | - | 2 - 3 | 433 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |