Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptlc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108406.1 | NP_001101876.1 | 1 - 2 | 72 | 124 |
Gene Symbol: Cyp3a23/3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | FQ218135.1 | - | 73 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |