Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kbtbd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107861.1 | NP_001101331.1 | 2 - 3 | 556 | 69 | |
XM_006236547.2 | XP_006236609.1 | 2 - 3 | 867 | 69 | ||
XM_006236548.2 | XP_006236610.1 | 2 - 3 | 711 | 69 | ||
XM_006236549.2 | XP_006236611.1 | 2 - 3 | 1128 | 69 | ||
XM_008762892.1 | XP_008761114.1 | 2 - 3 | 1233 | 69 | ||
XM_008762893.1 | XP_008761115.1 | 3 - 4 | 826 | 69 | ||
XM_008762894.1 | XP_008761116.1 | 2 - 3 | 601 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |