Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctdp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106131.1 | NP_001099601.1 | 11 - 12 | 2730 | 84 | |
XM_006254942.2 | XP_006255004.1 | 11 - 12 | 3835 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Protein Phosphatase Rattus norvegicus Probe spans exons |