Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyb5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022245.1 | NP_071581.1 | 1 - 2 | 200 | 68 | |
GenBank mRNA | AF007107.1 | - | 1 - 2 | 163 | 68 | |
AF007108.1 | - | 1 - 2 | 163 | 68 | ||
BC086945.1 | - | 1 - 2 | 194 | 7322141 | 68 | |
D13205.1 | - | 1 - 2 | 200 | 68 | ||
FQ209861.1 | - | 1 - 2 | 191 | 68 | ||
FQ210790.1 | - | 1 - 2 | 188 | 68 | ||
FQ224354.1 | - | 1 - 2 | 191 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |