Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pqlc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013189.1 | NP_001013207.1 | 1 - 2 | 321 | 73 | |
XM_006254952.2 | XP_006255014.1 | 2 - 3 | 613 | 73 | ||
XM_006254953.2 | XP_006255015.1 | 2 - 3 | 1006 | 73 | ||
XM_006254954.1 | XP_006255016.1 | 1 - 2 | 781 | 73 | ||
XM_006254955.2 | XP_006255017.1 | 2 - 3 | 612 | 73 | ||
XM_006254956.2 | XP_006255018.1 | 2 - 3 | 1003 | 73 | ||
XM_008772155.1 | XP_008770377.1 | 1 - 2 | 781 | 73 | ||
XM_008772156.1 | XP_008770378.1 | 1 - 2 | 781 | 73 | ||
GenBank mRNA | BC079072.1 | - | 1 - 2 | 779 | 73 | |
BC088186.1 | - | 1 - 2 | 321 | 7369419 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |