Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133519.4 | NP_598203.4 | 3 - 4 | 380 | 94 | |
GenBank mRNA | AF347935.1 | - | 3 - 4 | 397 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |