Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lilrb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013894.1 | NP_001013916.1 | 6 - 7 | 773 | 119 | |
XM_006228238.2 | XP_006228300.1 | 7 - 8 | 961 | 119 | ||
GenBank mRNA | AY359280.1 | - | 7 - 8 | 890 | 119 | |
AY359281.1 | - | 6 - 7 | 773 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |