Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277260.1 | NP_001264189.1 | 8 - 9 | 356 | 77 | |
NM_001277261.2 | NP_001264190.1 | 7 - 8 | 323 | 77 | ||
NM_001277262.2 | NP_001264191.1 | 7 - 8 | 326 | 77 | ||
NM_134388.2 | NP_599215.2 | 8 - 9 | 359 | 77 | ||
GenBank mRNA | AF399874.1 | - | 7 - 8 | 307 | 77 | |
AY334079.1 | - | 7 - 8 | 303 | 77 | ||
AY334080.1 | - | 7 - 8 | 300 | 77 | ||
AY334081.1 | - | 6 - 7 | 270 | 77 | ||
AY334082.1 | - | 6 - 7 | 267 | 77 | ||
FQ215948.1 | - | 6 - 7 | 356 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |