Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cab39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106924.1 | NP_001100394.1 | 1 - 2 | 372 | 146 | |
XM_006245373.2 | XP_006245435.1 | 1 - 2 | 402 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |