Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pax3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053710.1 | NP_446162.1 | 5 - 6 | 1178 | 56 | |
XM_006245131.2 | XP_006245193.1 | 5 - 6 | 1232 | 56 | ||
XM_008767181.1 | XP_008765403.1 | 6 - 7 | 1311 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |