Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhbdd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024891.1 | NP_001020062.1 | 2 - 3 | 688 | 80 | |
XM_008767250.1 | XP_008765472.1 | 2 - 3 | 1328 | 80 | ||
GenBank mRNA | BC097416.1 | - | 2 - 3 | 688 | 7454359 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |