Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019145.1 | NP_062018.1 | 10 - 11 | 1385 | 63 | |
GenBank mRNA | X74834.1 | - | 10 - 11 | 1385 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Detect Genomic DNA Rattus norvegicus Probe spans exons |