Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019145.1 | NP_062018.1 | 2 - 3 | 329 | 90 | |
GenBank mRNA | X74834.1 | - | 2 - 3 | 329 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Detect Genomic DNA Rattus norvegicus Probe spans exons |